Learn More
MP Biomedicals™ 1/4 in. Ceramic Spheres
Product Code. 11432400 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 Pk.
1000 Pk.
50 Pk.
500 Pk.
Unit Size:
Pack of 100
Pack of 1000
Pack of 50
Pack of 500
Product Code. 11432400
Brand: MP Biomedicals™ 116540422
Use to break hard/brittle samples. Specialty applications for the 1/4 in. Ceramic Spheres include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
  • Zirconium Oxide-coated ceramic grinding spheres
  • Chemically inert
  • Component of Lysing Matrix A and M tubes
TRUSTED_SUSTAINABILITY
Specifications
Bead Type Zirconium oxide coated ceramic
Content And Storage Room Temperature
Sample Type Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores
For Use With (Application) Break hard/brittle samples
Product Type Bulk Matrix
Quantity 1000 Pk.
Sample Volume Unlimited
Synonym 1/4 in. Zirconium Oxide Banded Satellite for Grinding
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.