Learn More
R&D Systems™ Mouse XL IL-15, Luminex Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Mouse |
| Gene ID (Entrez) | 3600 |
| Synonym | IL15, IL-15MGC9721, interleukin 15, interleukin-15 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |