Learn More
Mouse TIM-1/KIM-1/HAVCR Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 11 publications
Brand:  R&D Systems MAB1817-SP
 View more versions of this product
Product Code. 18745148
  • 209.00 EUR / 25µg
Description
TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TIM-1/KIM-1/HAVCR
Monoclonal
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
NP_001160104
HAVCR1
Mouse myeloma cell line NS0-derived recombinant mouse TIM-1/KIM-1/HAVCR Tyr22-Thr212 Accession # NP_001160104
25 μg
Primary
Detects mouse TIM-1/KIM-1/HAVCR in ELISAs and Western blots. This antibody does not cross-react with recombinant human (rh) TIM-1, rmTIM-2, or rmTIM-3.
Mouse
Purified
Western Blot, ELISA
222414
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
26762
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b
Certificates
Promotions
Promotions
For Research Use Only