missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse/Rat RGM-A Quantikine ELISA Kit

Product Code. 18772264
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18772264

Brand: R&D Systems™ MRGMA0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

a 4.5 hour solid phase ELISA designed to measure mouse and rat RGM-A in cell culture supernates, tissue homogenates, serum, and plasma.

The Mouse/Rat RGM-A Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat RGM-A in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (13 ul), edta plasma (13 ul), heparin plasma (13 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.08 - 5ng/mL
Assay Sensitivity 0.019ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and Recombinant Mouse and Rat RGM-A
For Use With (Application) ELISA
Gene ID (Entrez) 244058
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (13uL),EDTA Plasma (13uL),Heparin Plasma (13uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym RGM domain family member A, RGM domain family, member A, RGMA, RGMrepulsive guidance molecule A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.