missing translation for 'onlineSavingsMsg'
Learn More

Mouse RAGE Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18705446
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18705446 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18705446 Supplier R&D Systems Supplier No. FAB11795V100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

AGER Monoclonal specifically detects AGER in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAGE
Applications Flow Cytometry
Classification Monoclonal
Clone 697023
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.5 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_031451
Gene Alias advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1
Gene Symbols Ager
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Leu342 Accession # NP_031451
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5891
Test Specificity Detects mouse RAGE in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant canine RAGE and no cross-reactivity with recombinant human RAGE or recombinant rat RAGE is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.