missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse NGAL ELISA Kit

Product Code. 16861636 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Mouse
Unit Size:
96 tests
Product Code. Target Species unitSize
16861636 Mouse 96 tests
1 options
This item is not returnable. View return policy

Product Code. 16861636

Brand: Proteintech™ KE1004596T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KE10045 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The Mouse NGAL ELISA kit is to be used to detect and quantify protein levels of endogenous mouse NGAL.

  • Neutrophil gelatinase-associated lipocalin (NGAL; also known as neutrophil lipocalin, lipocalin-2, siderocalin, 24p3, or LCN2) is a small molecule of almost 25 kd that belongs to the well-defined superfamily of proteins called lipocalins.
  • NGAL initially found in activated neutrophils, in accordance with its role as an innate antibacterial factor. However, it subsequently was shown that many other types of cells, including in the kidney tubule, may produce NGAL in response to various injuries.
  • NGAL may become one of the most promising next-generation biomarkers in clinical nephrology and beyond.
  • The assay recognizes mouse NGAL. An antibody specific for mouse NGAL has been pre-coated onto the microwells.
  • The mouse NGAL protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for mouse NGAL is added to detect the captured mouse NGAL protein.
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent.
  • Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.

Specifications

Accession Number BC132071
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 4 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Mouse
Gene ID (Entrez) 16819
Host Species Mouse
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P11672
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Mouse NGAL ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.