Learn More
Lonza™ Mouse Neural Stem Cell Nucleofector™ Kit
Description
- High transfection efficiencies for primary cells.
- Cells maintain functionality post transfection.
- Applications:
- Transfection of cell numbers in the range of 2 × 105 to 2 × 107 cells.
- Low-throughput transfection in a single cuvette.
Specifications
Specifications
| Color | Clear |
| Vector | pmaxGFP™ |
| For Use With (Equipment) | Nucleofector™ I/II/2b Device |
| For Use With (Application) | Efficient transfection of primary mouse neural stem cells |
| Includes | 2.25 mL Nucleofector™ Solution for Mouse Neural Stem Cells, 0.5 mL Supplement 1, 30 μg pmaxGFP™ Vector (0.5 μg/μL in 10 mM Tris pH 8.0), 25 x Aluminum cuvettes (100 μL), 25 x Single-use pipettes |
| No. of Reactions | 25 |
| pH | 8 |
| Quantity | 1 Kit |