missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Mouse Il7 (P10168) Recombinant Protein

Product Code. 16121610
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 16121610

Brand: Abnova™ P4583.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Sequence: MECHIKDKEGKAYESVLMISIDELDKMTGTDSNCPNNEPNFFRKHVCDDTKEAAFLNRAARKLKQFLKMNISEEFNVHLLTVSQGTQTLVNCTSKEEKNVKEQKKNDACFLKRLLREIKTCWNKILKGSI

Specifications

Accession Number P10168
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 16196
Molecular Weight (g/mol) 17.4kDa
Name Il7 (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Immunogen MECHIKDKEGKAYESVLMISIDELDKMTGTDSNCPNNEPNFFRKHVCDDTKEAAFLNRAARKLKQFLKMNISEEFNVHLLTVSQGTQTLVNCTSKEEKNVKEQKKNDACFLKRLLREIKTCWNKILKGSI
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias A630026I06Rik/Il-7/MGC129342/hlb368
Common Name Il7
Gene Symbol Il7
Biological Activity The activity is determined by dose-dependent induction of 2E8 cell proliferation and is typically less than 0.2ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.