missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-28A/B (IFN-lambda 2/3) Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
209.00 EUR - 2060.00 EUR
Specifications
| Antigen | IL-28A/B (IFN-lambda 2/3) |
|---|---|
| Clone | 244716 |
| Concentration | LYOPH |
| Dilution | Neutralization 3-9 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721137
![]() |
R&D Systems
MAB17892-SP |
25 μg |
209.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18710667
![]() |
R&D Systems
MAB17892-100 |
100 μg |
512.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701917
![]() |
R&D Systems
MAB17892-500 |
500 μg |
2060.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-28A/B (IFN-lambda 2/3) Monoclonal specifically detects IL-28A/B (IFN-lambda 2/3) in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| IL-28A/B (IFN-lambda 2/3) | |
| LYOPH | |
| Neutralization, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8CGK6 | |
| 282616 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 244716 | |
| Neutralization 3-9 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IFNl2, IFNlambda 2, IFN-lambda 2, IFN-lambda 3, IL28A, IL-28A, IL28B, interferon lambda 2 | |
| E. coli-derived recombinant mouse IL-28B/IFN-lambda 3 Asp20-Val193 Accession # Q8CGK6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-28B/IFN-lambda 3 in ELISAs. In sandwich immunoassays, 100% cross-reactivity with recombinant mouse IL-28A/IFN-lambda 2 and no cross-reactivity with recombinant human (rh) IL-28A or rhIL-29 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title