missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-gamma APC-conjugated Antibody, R&D Systems™

Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
Specifications
| Antigen | IFN-gamma |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 37895 |
| Conjugate | APC |
| Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Alias | IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma |
| Gene Symbols | IFNG |
| Host Species | Rat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?