Learn More
Mouse HB-EGF Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB8239-SP
 View more versions of this product
Product Code. 18775109
  • 231.00 EUR / 25µg
Description
HB-EGF Monoclonal specifically detects HB-EGF in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HB-EGF
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), Dtr, DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), Dts, DTSF, HBEGF, Hegfl, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor
E. coli-derived recombinant mouse HB-EGF Asp63-Leu148 Accession # Q06186
25 μg
1839
Reconstitute at 0.5mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
ELISA
917525R
ELISA
Q06186
Rat
Protein A or G purified from cell culture supernatant
Primary
Detects mouse HB-EGF in direct ELISAs.
Mouse
Supernatant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions