missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Glut2 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB1440
This item is not returnable.
View return policy
Description
Glut2 Monoclonal specifically detects Glut2 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Glut2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P14246 | |
| SLC2A2 | |
| CHO Chinese hamster ovary cell line transfected with mouse Glut2 Met1-Val523 Accession # P14246 | |
| 200 μg | |
| Primary | |
| Detects mouse Glut2. Recognizes mouse Glut2-transfected NS0 cells but not NS0 control transfectants. It also detects Glut2 on mouse insulinoma beta TC-6 cells (2). | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 205115 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6514 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction