missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Epigen ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Mouse Epigen quantitates mouse Epigen in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Epigen.
Epigen is an EGF-related polypeptide growth factor that signals through the ErbB receptor-1. Promotes the growth of epithelial cells. May stimulate the phosphorylation of EGFR and mitogen-activated protein kinases. It is produced in several tissues, including the testis, liver, and heart, as well as in certain tumor cells. Epigen is mitogenic for fibroblasts and epithelial cells. Human Epigen is initially synthesized as a glycosylated 14.7 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a mature soluble sequence.
Specifications
Specifications
| Accession Number | 2310069M11Rik; ALGV3072; EPG; EPGN; epigen; Epithelial mitogen; epithelial mitogen homolog; PRO9904; RGD1560084; UNQ3072/PRO9904 |
| Assay Range | 6.2 to 1,500 pg/mL |
| Assay Sensitivity | 6.2 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 71920 |
| Gene Symbol | EPGN |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction