missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse Cytokine Positive Control Cells

Product Code. 15596266
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15596266 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15596266 Supplier Invitrogen™ Supplier No. 00450051

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recommended for use as positive control cells when establishing an intracellular cytokine staining protocol.

  • Mouse splenocytes were stimulated with ConA (3μg/mL) for two days, followed by recombinant IL-2 (20ng/mL) and IL-4 (20ng/mL) for three days
  • The cells were then restimulated with immobilized anti-mouse CD3 (10μg/mL) and soluble anti-mouse CD28 (2μg/mL) in the presence of Brefeldin A for 5 hours
  • These cells contain subpopulations that express IL-2, IL-3, IL-4, IL-10, GM-CSF, and IFNγ
  • The cells have been fixed with formaldehyde and are provided in a stabilizing buffer so that they may be stored at −80°C for up to six months or at 2 to 8°C for up to one month
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Flow Cytometer
Quantity 1 mL
Product Line eBioscience
Product Type Mouse Cytokine Positive Control Cells

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.