missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cystatin C Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1238
This item is not returnable.
View return policy
Description
Cystatin C Polyclonal specifically detects Cystatin C in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, ELISA Capture (Matched Antibody Pair).
Specifications
| Cystatin C | |
| Polyclonal | |
| Unconjugated | |
| P21460 | |
| CST3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cystatin C (R & D Systems, Catalog # 1238-PI) Ala21-Ala140 Accession # P21460 | |
| 100 μg | |
| Primary | |
| Detects mouse and human Cystatin C in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human Cystatin D is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| ARMD11, bA218C14.4 (cystatin C), CST3, cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, Post-gamma-globulin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1471 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction