missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL3/MIP-1 alpha Antibody, R&D Systems™

Rat Monoclonal Antibody
209.00 EUR - 429.00 EUR
Specifications
| Antigen | CCL3/MIP-1 alpha |
|---|---|
| Clone | 756605 |
| Dilution | Neutralization 0.125-0.75 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18708179
![]() |
R&D Systems
MAB4502-SP |
25 μg |
209.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18748179
![]() |
R&D Systems
MAB4502-100 |
100 μg |
429.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Mouse samples. It is validated for Neutralization.Specifications
| CCL3/MIP-1 alpha | |
| Neutralization 0.125-0.75 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
| CCL3 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 756605 | |
| Neutralization | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P10855 | |
| 6348 | |
| E. coli-derived recombinant mouse CCL3/MIP-1 alpha Ala24-Ala92 Accession # P10855 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CCL3/MIP-1 alpha in direct ELISAs. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title