missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse Anti-SARS-CoV-2 Antibody IgG Titer Serologic Assay kit
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
The ongoing SARS-COV-2 pandemics has sickened 3.7 million people around the world, claiming over 1,000,000 lives. To deal with this global public health crisis, unprecedented efforts are being made to study the virus, SARS-CoV-2. At present, there are 193 new crown vaccine projects registered with the World Health Organization are being developed. To support these studies, ACRO has developed Anti-SARS-CoV-2 subtype antibody Serologic Assay kit including:IgG1, IgG2 IgG3, IgG4.
Specifications
Specifications
| Product Type | Antibody titer assay kits |
| Kit Contents | This assay kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 antibodies in Mouse serum by SARS-CoV-2 Spike RBD. The Kit consists of Pre-coated with SARS-CoV-2 Spike RBD Microplate, an Anti-SARS-CoV-2 Antibody (C) |
| Quantity | 96 Tests |
| Shipping Condition | RT |
| Storage Requirements | 2°C to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction