missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rabbit IgM (H+L) Secondary Antibody (NRBM), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68467MFV450
This item is not returnable.
View return policy
Description
IgM (H+L) Monoclonal antibody specifically detects IgM (H+L) in Rabbit samples. It is validated for Flow Cytometry
Specifications
| IgM (H+L) | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit IgM | |
| 0.1 mL | |
| Secondary | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| NRBM | |
| Flow Cytometry | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction