missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRL, Mouse, Clone: ME.122, Abnova™
Mouse monoclonal antibody raised against PRL.
Brand: Abnova MAB4303.1mg
This item is not returnable.
View return policy
Specifications
| PRL | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PRL | |
| Mouse | |
| 1 mg | |
| Primary | |
| Recognizes the beta subunit of human prolactin. Specificity is tested by EIA. | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| ELISA | |
| ME.122 | |
| Mouse monoclonal antibody raised against PRL. | |
| In 150mM PBS, pH 7.4 | |
| PRL | |
| Human Prolactin. | |
| RUO | |
| 5617 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction