missing translation for 'onlineSavingsMsg'
Learn More

Mosapride citrate, MedChemExpress

Product Code. p-200099913
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16438398

Brand: Medchem Express HYB0189A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mosapride citrate is a gastroprokinetic agent that acts as a selective 5HT4 agonist.

Mosapride citrate is a gastroprokinetic agent that acts as a selective 5HT4 agonist.

Specifications

Chemical Name or Material Mosapride citrate
CAS 112885-42-4
Color White
Purity Grade Notes Research
Molecular Formula C27H33ClFN3O10
Quantity 10 mg
Synonym TAK-370 citrate AS-4370 citrate
Solubility Information DMSO : 100 mg/mL (162.86 mM; Need ultrasonic)
SMILES O=C(NCC1CN(CC2=CC=C(F)C=C2)CCO1)C3=CC(Cl)=C(N)C=C3OCC.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 614.02
Formula Weight 614.02
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.