missing translation for 'onlineSavingsMsg'
Learn More

(Morpholinium-4-ylmethyl)trifluoroborate internal salt, 95%

Product Code. p-7031897
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15478335

Brand: Thermo Scientific Alfa Aesar H33149.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1268340-94-8 | C5H11BF3NO | 168.954 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1268340-94-8
Molecular Formula C5H11BF3NO
Molecular Weight (g/mol) 168.954
MDL Number MFCD20134168
InChI Key FQKLIGKQQGNVHW-UHFFFAOYSA-O
Synonym trifluoro morpholin-4-ium-4-ylmethyl boranuide, morpholinium-4-ylmethyl trifluoroborate internal salt, morpholinium-4-yl-methyl trifluoroborate internal salt, morpholinium-4-ylmethyltrifluoroboronanion, morpholin-4-ium-4-yl methyl trifluoroborate, trifluoro morpholin-4-ium-4-yl methyl boranuide, trifluoro morpholin-4-ium-4-yl methyl borate 1-
PubChem CID 53243644
IUPAC Name trifluoro(morpholin-4-ium-4-ylmethyl)boranuide
SMILES [B-](C[NH+]1CCOCC1)(F)(F)F

Specifications

Melting Point 170°C to 172°C
Quantity 1 g
Formula Weight 168.95
Percent Purity 95%
Chemical Name or Material (Morpholinium-4-ylmethyl)trifluoroborate internal salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.