missing translation for 'onlineSavingsMsg'
Learn More

Morpholine-4-carbothioamide, 97%, Thermo Scientific™

Product Code. p-3486256
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295604 250 mg Amber Glass Bottle 250mg
10492444 1 g Amber Glass Bottle 1g
10125343 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10295604 Supplier Thermo Scientific Maybridge Supplier No. RB00140CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 14294-10-1
Molecular Formula C5H10N2OS
Molecular Weight (g/mol) 146.21
MDL Number MFCD00178434
InChI Key GSLBUBZXFUYMSW-UHFFFAOYSA-N
Synonym 4-morpholinecarbothioamide, 1-morpholinethiocarboximide, 4-morpholinethiocarboxamide, morpholine-4-carbothioic acid amide, aminomorpholin-4-ylmethane-1-thione, thiocarbamoylmorpholine, morpholine-4-carbothioicacidamide, 4-amino thiocarbonyl morpholine
PubChem CID 2393544
IUPAC Name morpholine-4-carbothioamide
SMILES NC(=S)N1CCOCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 146.21
Percent Purity 97%
Chemical Name or Material Morpholine-4-carbothioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.