missing translation for 'onlineSavingsMsg'
Learn More

Morinidazole (R enantiomer), MedChemExpress

Product Code. p-200102214
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469377 5 mg 5mg
16426838 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469377 Supplier Medchem Express Supplier No. HY15781A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Morinidazole R enantiomer is the R-enantiomer of Morinidazole. Morinidazole is a new 5-nitroimidazole class antimicrobial agent. Morinidazole R enantiomer is the less active enantiomer.

Morinidazole R enantiomer is the R-enantiomer of Morinidazole. Morinidazole is a new 5-nitroimidazole class antimicrobial agent. Morinidazole R enantiomer is the less active enantiomer.

Specifications

Chemical Name or Material Morinidazole (R enantiomer)
Physical Form Solid
CAS 898230-59-6
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H18N4O4
For Use With (Application) COVID-19-immunoregulation
Synonym R-Morinidazole
Solubility Information DMSO : ≥ 100 mg/mL (369.97 mM)
SMILES O[C@@H](CN1C([N+]([O-])=O)=CN=C1C)CN2CCOCC2
Molecular Weight (g/mol) 270.29
Formula Weight 270.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.