Learn More
Mordant Black, Fisher Chemical™
Product Code. 12970464 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
Unit Size:
100mL
Product Code. 12970464
Brand: Fisher Chemical RP 130
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Color Black
Quantity 100 g
Product Identifier
  • Mordant black 11 triturate. European Pharmacopoeia Ref: 1056801.