missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MORC1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98518PCP
This item is not returnable.
View return policy
Description
MORC1 Polyclonal antibody specifically detects MORC1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MORC1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 27136 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PerCP | |
| CT33microrchidia homolog (mouse), microrchidia (mouse) homolog, MORC, MORC family CW-type zinc finger 1, MORC family CW-type zinc finger protein 1, mouse, homolog of, ZCW6 | |
| Produced in rabbits immunized with E. coli-derived Human MORC1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction