Learn More
Monofax 057 Surfactant, Pure, (Mono And Di-Phosphate Esters), Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
500mL
Specifications
Specifications
| Color | Yellow |
| pH | 1.3 |
| Boiling Point | 93.33°C |
| Packaging | Plastic Coated Glass bottle |
| MDL Number | 283882 |
| Quantity | 500 mL |
| Physical Form | Liquid |