Learn More
Mono[2-(perfluorohexyl)ethyl] Phosphate Dibenzyl Ester, TRC
Product Code. 30480148
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480148 250 mg 250mg
30479236 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30480148 Supplier Toronto Research Chemicals Supplier No. TRCM566805250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Chemical Name or Material Mono[2-(perfluorohexyl)ethyl] Phosphate Dibenzyl Ester
Analyte or Component Names Mono[2-(perfluorohexyl)ethyl] Phosphate Dibenzyl Ester
Shipping Condition Room Temperature
Molecular Formula C22 H18 F13 O4 P
Synonym Dibenzyl (3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl) Phosphate
InChI Formula InChI=1S/C22H18F13O4P/c23-17(24,18(25,26)19(27,28)20(29,30)21(31,32)22(33,34)35)11-12-37-40(36,38-13-15-7-3-1-4-8-15)39-14-16-9-5-2-6-10-16/h1-10H,11-14H2
Solution Type Neat
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOP(=O)(OCc1ccccc1)OCc2ccccc2
IUPAC Name dibenzyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl phosphate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 624.329
Quantity 250 mg
Formula Weight 624.074
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.