missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus E8L Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
213.00 EUR - 507.00 EUR
Specifications
| Antigen | Monkeypox Virus E8L |
|---|---|
| Dilution | Western Blot 1:2000 - 1:6000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231638
|
Novus Biologicals
NBP3-33396-100ul |
100 μL |
507.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226532
|
Novus Biologicals
NBP3-33396-20ul |
20 μL |
213.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Monkeypox Virus E8L Monoclonal antibody specifically detects Monkeypox Virus E8L in Virus samples. It is validated for ELISA,Western BlotSpecifications
| Monkeypox Virus E8L | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of E8L. (NP_536532.1).,, Sequence:, MPQQLSPINIETKKAISDTRLKTLDIHYNESKPTTIQNTGKLVRINFKGGYISGGFLPNEYVLSTIHIYWGKEDDYGSNHLIDVYKYSGEINLVHWNKKKYSSYEEAKKHDDGIIIIAIFLQVSDHKNVYFQKIVNQLDSIRSANMSAPFDSVFYLDNLLPSTLDYFTYLGTTINHSADAAWIIFPTPINIHSDQLSKFRTLLSSSNHEGKPHYITENYRNPYKLNDDTQVYYSGEIIRAATTSPVRENYFMKWLSDLREACFSYYQKYIEGNKT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:2000 - 1:6000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title