missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkey GASP-1 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Monkey GASP-1 quantitates monkey GASP-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey GASP-1.
Modulates lysosomal sorting and functional down-regulation of a variety of G-protein coupled receptors. Targets receptors for degradation in lysosomes via its interaction with BECN2.
Specifications
Specifications
| Accession Number | 2210415K24Rik; 3110031O14Rik; C87852; G protein-coupled receptor associated sorting protein 1; GASP; GASP1; GASP-1; GPRASP1; G-protein coupled receptor-associated sorting protein 1; Kiaa0443; Per1 interacting protein; per1-interacting protein; Pips |
| Assay Range | 49 to 12,000 pg/mL |
| Assay Sensitivity | 49 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Interassay CV | <12% |
| Intraassay CV | <10 |
| Kit Contents | Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction