missing translation for 'onlineSavingsMsg'
Learn More

Molibresib, MedChemExpress

Product Code. 16408217
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408217 5 mg 5mg
16476076 100 mg 100mg
16465498 10 mg 10mg
16431066 200 mg 200mg
16421677 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16408217 Supplier Medchem Express Supplier No. HY13032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Molibresib (I-BET762; GSK525762) is a BET bromodomain inhibitor with IC50 of 32.5-42.5 nM.

Molibresib (I-BET762

Specifications

Chemical Name or Material Molibresib
CAS 1260907-17-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H22ClN5O2
Quantity 5 mg
Synonym I-BET762 GSK525762 GSK525762A
Solubility Information DMSO : 200 mg/mL (471.81 mM; Need ultrasonic)
SMILES ClC1=CC=C(C2=N[C@@H](CC(NCC)=O)C3=NN=C(C)N3C4=CC=C(OC)C=C24)C=C1
Molecular Weight (g/mol) 423.9
Formula Weight 423.9
Percent Purity 98.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.