Learn More
Molecular Sieves, 5Å, Honeywell Fluka™
Product Code. p-7120600 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with Inliner
Poly Bottle
Unit Size:
1kg
5kg
Product Code. 15661850
Brand: Honeywell-Fluka 2086201KG
Request Bulk or Custom Format
Beads, 8-12 mesh
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Specifications
Mesh Size 8 to 12mesh
Packaging Poly Bottle
Pore Size 5 Å
Quantity 1 kg
Physical Form Beads
Linear Formula Ca/nNa12-2n[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
IUPAC Name Molecular sieves 5A
Chemical Name or Material Molecular Sieves, 5Å