Learn More
Invitrogen™ SYTO™ 85 Orange Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO
Description
The cell-permeant SYTO 85 orange fluorescent nucleic acid stain exhibits bright, orange fluorescence upon binding to nucleic acids. Because the staining pattern of the SYTO dyes in live cells may vary between cell types, we offer the SYTO Orange Fluorescent Nucleic Acid Stain Sampler Kit (Cat. No. S-11360) to enable researchers to find the most appropriate orange-fluorescent SYTO stain for their system.
Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Orange |
| Content And Storage | Store in freezer at -5°C to -30°C and protect from light. |
| Excitation Wavelength Range | 567 nm |
| Dye Type | Cell-Permeant |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 250 μL |
| Volume (Metric) | 250 μL |
| Detection Method | Fluorescence |
| Description | SYTO™ 85 Orange Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO |
| Emission | 583 nm |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.