missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Qtracker™ 655 Vascular Labels
Click to view available options
Quantity:
200 μL
Unit Size:
Each
Description
Includes
200μL Qtracker non-targeted quantum dots (2μM solution in 50mm borate buffer, pH 8.3).
- These nanocrystals exhibit intense fluorescence with red-shifted emission for increased tissue penetration, and have a PEG surface coating specially developed to minimize nonspecific interactions and reduce immune response by the tissue
- Because PEG surface coating does not contain reactive functional groups, dots are retained in circulation longer and can be imaged for up to 3 hours with single injection or for longer periods of time with additional injections
- Qtracker 655 label has Ex/Em (405-615/655)nm
Cell Analysis, Cellular Imaging, In Vivo Imaging
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Concentration | 2 μM |
| Content And Storage | Contains 200 μl Qtracker™ non-targeted quantum dots (2 μM solution in 50 mM borate buffer, pH 8.3). Store at 2–6°C. Do not freeze. Stable for at least 6 months. |
| Dye Type | Qdot Nanocrystals |
| Quantity | 200 μL |
| Product Line | QTRACKER, Qdot |
| Reagent Type | Vascular Imaging Reagents |
| Shipping Condition | Room Temperature |
| Technique | In Vivo Imaging |
| Product Type | Vascular Label |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction