Learn More
Invitrogen™ Ovalbumin, Alexa Fluor™ 647 Conjugate
Click to view available options
Quantity:
2 mg
Unit Size:
Each
Description
The far-red fluorescent Alexa Fluor 647 ovalbumin conjugate is a protein with a relatively low molecular weight (∽45,000 daltons).
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, In Vivo Imaging
Informations sur la commande
Shipping Condition: Room Temperature
Spécification
Spécification
| Quantity | 2 mg |
| Conjugate | Alexa Fluor 647 |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Albumin Proteins |
| Product Line | Alexa Fluor™ |
For Research Use Only. Not for use in diagnostic procedures.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu