Learn More
Molecular Probes™ Escherichia coli (K-12 strain) BioParticles , BODIPY™ FL conjugate
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
- Conjugates covalently labeled with a variety of different fluorophores (special care has been taken to remove free dye after conjugation)
- Provided as lyophilized powders
- Approximately 3 x 108 E. coli or S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid
- Conjugates can be reconstituted in buffer of choice for use in phagocytosis assays
- Fluorescence of conjugates that are bound to surface ofcell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers
- In addition to cellular applications, fluorescent conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants
- Small particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways
Cell Culture, Mammalian Cell Culture, Neuronal Cell Culture, Primary Cell Culture
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 10 mg |