missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CoroNa™ Green, AM, cell permeant - Special Packaging

Product Code. 11544277
Click to view available options
Quantity:
20 x 50 μg
Unit Size:
1000µg
This item is not returnable. View return policy

Product Code. 11544277

Brand: Invitrogen™ C36676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A sodium ion indicator that exhibits an increase in green fluorescence emission intensity upon binding Na+, with little shift in wavelength

  • Allows spatial and temporal resolution of Na+ concentrations in presence of physiological concentrations of other monovalent cations
  • Dye loads into cells more efficiently than does sodium green tetraacetate, and CoroNa Green indicator responds to broader range of Na+ concentration
  • With absorption/emission maxima of approximately 492/516nm for Na+-bound form, can be detected by any instrument that can detect fluorescein

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Sodium and Potassium Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type Sodium Indicator
Dye Type Sodium Indicator
Product Line CoroNa
Quantity 20 x 50 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.