Learn More
Invitrogen™ CellLight™ Null (control), BacMam 2.0
Description
CellLight Null Control contains no mammalian promoter and no transgene. CellLight reagents combine the utility and selectivity of fluorescent proteins with the transduction efficiency of BacMam technology, enabling unambiguous staining of organelles, cellular structures, and processes in live mammalian cells. They are provided in a ready-to-use format—simply add, incubate, and image—with highly efficient expression in cell lines, primary cells, stem cells, and neurons.Other CellLight reagents are fusion constructs of signal peptides or cell structure proteins with premiere emGFP, TagRFP or CFP for accurate and specific targeting to sub-cellular compartments and structures. A variety of targets, including cytoskeleton, mitochondria, and secretory compartments, are available for convenient multiplexing, co-localization studies, and imaging of dynamic cellular processes where high spatial and temporal resolution is required. The CellLight reagents tolerate fixation with formaldehyde and are therefore compatible with fixed-cell analysis.
- Highly Efficient: >90% transduction of wide range of mammalian cell lines, including primary cells, stem cells, and neurons
- Fast and Convenient: Simply add CellLight Null Control reagent to cells, incubate overnight, and evaluate how well cells tolerate BacMam transduction
- Safe: CellLight reagents are non-replicating in mammalian cells, lack of observable cytopathic effect, and are suitable for biosafety level (BSL) 1 handling
Cell Analysis, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Color | Colorless |
| Content And Storage | Store at 2°C to 6°C, protected from light. Do Not Freeze. |
| Dye Type | Unconjugated |
| For Use With (Equipment) | Confocal Microscope, Fluorescence Microscope |
| Quantity | 1 mL |
| Detection Method | Fluorescence |
| Emission | Visible |
| Form | Liquid |
| Product Line | CellLight |
| Shipping Condition | Wet Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.