missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Albumin from Bovine Serum (BSA), Alexa Fluor™ 594 conjugate

Product Code. 11598926
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 11598926

Brand: Invitrogen™ A13101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bright and photostable conjugate exceptionally suited for applications in which well-defined molecular weight tracers are needed

  • Applications include studying rate of endocytosis and exocytosis and following intracellular protein processing

Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Conjugate Alexa Fluor 594
Species Bovine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Albumin Proteins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.