Learn More
Thermo Scientific™ Molecular BioProducts™ HLT Ergo-F Pipet Tips
GREENER_CHOICE
Product Code. 12924077 Shop All MBP Molecular BioProducts Products
Click to view available options
Volume (Metric):
200 μL
Unit Size:
Pack of 960
Product Code. 12924077
Brand: Thermo Scientific™ 3812
Only null left
Add to basket
Add to basket
For use with Finnpipette Focus and Digital Pipetters
  • Ergonomic tips reduce strains and injury
  • Secure and consistent seals
  • Less ejection force
  • Also available in low retention polymer
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 μL
Sterility Non-sterile
Format Rack
For Use With (Equipment) Thermo Scientific Finnpipette Focus, Digital Pipette
Product Line Molecular BioProducts HLT Ergo-F