Learn More
DuPont™ Tychem 6000 F CHA5 Hooded Coverall
Offering excellent chemical permeation protection to an extensive range of chemicals. DuPont™ Tychem™ 6000 F Hooded Coveralls are made from a lightweight and durable fabric consisting of a proprietary barrier film-laminated to a heavy duty Tyvek substrate.
36.80 EUR - 895.00 EUR
Specifications
Product Type | Hooded Coveralls |
---|---|
Material | Tychem |
Color | Gray |
Waist Style | Elastic |
Ankle Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11793083
|
DuPont™
D13395221 |
Large |
885.00 EUR
Pack of 25 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12309429
|
DuPont™
D13395221 |
Large |
36.80 EUR
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12623680
|
DuPont™
D13495186 |
Small |
885.00 EUR
Pack of 25 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
11703093
|
DuPont™
D13395545 |
X-Large |
885.00 EUR
Pack of 25 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12325659
|
DuPont™
D13395545 |
X-Large |
36.80 EUR
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
11713093
|
DuPont™
D13395268 |
2X-Large |
885.00 EUR
Pack of 25 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12399419
|
DuPont™
D13395268 |
2X-Large |
36.80 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
12319429
|
DuPont™
D13495156 |
Medium |
36.80 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
12613680
|
DuPont™
D13495156 |
Medium |
885.00 EUR
Pack of 25 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12603680
|
DuPont™
D13495118 |
3X-Large |
895.00 EUR
Pack of 25 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
- Available in grey for discretionary needs
- Elasticated face, wrists, waist and ankles for optimal fit
- Thumb loops prevent sleeves from riding up
- Self-adhesive chin flap for tight seal of suit to face masks
- Self-adhesive double zipper flap closure system
- Stitched and over-taped seams with barrier tape for protection and strength
- Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
- Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
Specifications
Hooded Coveralls | |
Gray | |
Elastic | |
Hooded | |
Double Zipper with Self-adhesive Flap | |
Category III, Type 3-B, 4-B, 5-B and 6-B | |
Self-adhesive chin flap | |
For a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications |
Tychem | |
Elastic | |
Elastic | |
Elastic | |
Stitched, Taped | |
Chemicals | |
(EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5 |