missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOC-POC Tenofovir Fumarate Salt (Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | MOC-POC Tenofovir Fumarate Salt (mixture of diastereomers) |
| Molecular Formula | C17 H26 N5 O10 P . C4 H4 O4 |
| Quantity | 10 mg |
| Synonym | 5-[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]-2,4,6,8-tetraoxa-5-phosphanonanedioic Acid (1-Methylethyl,methyl) Ester 5-Oxide Fumarate, MOC-POC PMPA Fumarate |
| InChI Formula | InChI=1S/C17H26N5O10P.C4H4O4/c1-11(2)32-17(24)28-9-31-33(25,30-8-27-16(23)26-4)10-29-12(3)5-22-7-21-13-14(18)19-6-20-15(13)22;5-3(6)1-2-4(7)8/h6-7,11-12H,5,8-10H2,1-4H3,(H2,18,19,20);1-2H,(H,5,6)(H,7,8)/b;2-1+/t12-,33?;/m1./s1 |
| SMILES | COC(=O)OCOP(=O)(CO[C@H](C)Cn1cnc2c(N)ncnc12)OCOC(=O)OC(C)C.OC(=O)\C=C\C(=O)O |
| IUPAC Name | [[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-(methoxycarbonyloxymethoxy)phosphoryl]oxymethyl propan-2-yl carbonate;(E)-but-2-enedioic acid |
| Molecular Weight (g/mol) | 607.46 |
| Formula Weight | 607.1527 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?