Learn More
Miele Professional™ Mobile Injector
Product Code. 10683191 Shop All Miele Products
Click to view available options
:
Holds: 38 Pipettes
Product Code. 10683191
Brand: Miele Professional™ 6727920
This item requires a price update prior to being ordered.
Request a Price for this item here.
Mobile unit for optimum loading of pipettes
  • Equipped with 38 pipettes in 3 rows
  • 1st Row: 10 pipettes x 100 ml (up to 550 mm), bar distance 20 mm
  • 2nd Row: 14 pipettes x 25 ml , bar distance 26 mm
  • 3rd Row: 14 pipettes x 10 ml, bar distance 26 mm
  • Bracket for ML/2 for automatic mobile unit recognition
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mobile Injector Unit with Drying Connection
Model E 405/1
For Use With (Equipment) Pipettes
Quantity 1 Each