Description
- Equipped with 6 nozzles
- Equipped with a height-adjustable holding frame
- Maximum vertical clearance from star overlay: 610 mm
Specifications
Specifications
| Material | Stainless Steel, Plastic |
| Color | Stainless Steel, Black |
| Holds | 6 Large volume laboratory glassware |
| For Use With (Equipment) | Washer-disinfectors, laboratory |
| Depth (Metric) | 600 mm |
| Height (Metric) | 346 mm |
| Width (Metric) | 530 mm |