missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MOB3A/MOB3B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16627714
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16627714 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16627714 Supplier Invitrogen™ Supplier No. PA5101233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MOBKL2A/B.

MOBKL2A (Mps one binder kinase activator-like 2A), also known as MOB-LAK or MOB3A, is a 217 amino acid protein that regulates kinase activity. A member of the MOB1/phocein family, MOBKL2A is encoded by a gene that maps to human chromosome 19. Consisting of around 63 million bases with over 1,400 genes, chromosome 19 makes up over 2% of human genomic DNA. Chromosome 19 includes a diversity of interesting genes and is recognized for having the greatest gene density of the human chromosomes. It is the genetic home for a number of immunoglobulin superfamily members including the killer cell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAM and PSG family, and Fcα receptors. Key genes for eye color and hair color also map to chromosome 19. Peutz-Jeghers syndrome, spinocerebellar ataxia type 6, the stroke disorder CADASIL, hypercholesterolemia and insulin-dependent diabetes have been linked to chromosome 19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOB3A/MOB3B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MOB3B
Gene Accession No. Q86TA1, Q8BSU7, Q8VE04, Q96BX8
Gene Alias 5330417K06Rik; 8430436F23Rik; A430018A01Rik; A630029F06; AV218468; C9orf35; MOB kinase activator 3A; MOB kinase activator 3B; MOB kinase activator-like 2B; MOB LAK; MOB1 (Mps One Binder) kinase activator-like 2B; mob1 homolog 2A; Mob1 homolog 2b; MOB1, Mps One Binder kinase activator-like 2A; MOB1, Mps One Binder kinase activator-like 2B; MOB1C; MOB1D; Mob3a; MOB3B; MOBKL2A; Mobkl2b; moblak; MOB-LAK; monopolar spindle 1 binding, MOB1, domain containing; mps one binder kinase activator-like 2A; Mps one binder kinase activator-like 2B; RGD1560696
Gene Symbols MOB3A, MOB3B
Host Species Rabbit
Immunogen A synthesized peptide derived from human MOB3A(Accession Q96BX8), corresponding to amino acid residues M1-N40.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126308, 208228, 214944, 79817
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.