Learn More
MMP12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22989-1-AP-150UL
 View more versions of this product
Product Code. 16812155
  • 410.00 EUR / 150µL
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. It is thought that the protein encoded by this gene is cleaved at both ends to yield the active enzyme, but this processing has not been fully described. The enzyme degrades soluble and insoluble elastin. It may play a role in aneurysm formation and studies in mice suggest a role in the development of emphysema. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3.
Specifications
Specifications
MMP12
Polyclonal
Unconjugated
MMP12
HME, Macrophage elastase, Macrophage metalloelastase, Matrix metalloproteinase 12, ME, MME, MMP 12, MMP12
Rabbit
Antigen Affinity Chromatography
RUO
17381, 4321
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.52 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P34960, P39900
Mmp12
MMP12 Fusion Protein Ag19068
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions