missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-25/MT6-MMP Antibody (141811), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB11422W
This item is not returnable.
View return policy
Description
MMP-25/MT6-MMP Monoclonal antibody specifically detects MMP-25/MT6-MMP in Human samples. It is validated for Flow Cytometry
Specifications
| MMP-25/MT6-MMP | |
| Monoclonal | |
| DyLight 650 | |
| leukolysin, matrix metallopeptidase 25, matrix metallopeptidase-like 1, matrix metalloproteinase 20, matrix metalloproteinase 25, matrix metalloproteinase-25, matrix metalloproteinase-like 1, membrane-type 6 matrix metalloproteinase, membrane-type matrix metalloproteinase 6, membrane-type-6 matrix metalloproteinase, MMP20, MMP20A, MMP-25, MMPL1, MT6MMP, MT6-MMP, MT-MMP 6, MTMMP6, MT-MMP6 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-25/MT6-MMP, Pro314-Cys508, Accession # Q9NPA2 | |
| 0.5 mL | |
| Angiogenesis, Cancer | |
| 64386 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 141811 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction