missing translation for 'onlineSavingsMsg'
Learn More

MM-102 TFA, MedChemExpress

Product Code. 16467757
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16467757

Brand: Medchem Express HY12220A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MM-102 TFA (HMTase Inhibitor IX TFA) is a potent WDR5/MLL interaction inhibitor, achieves IC50 = 2.4 nM with an estimated Ki 200 times more potent than the ARA peptide.

MM-102 TFA (HMTase Inhibitor IX TFA) is a potent WDR5/MLL interaction inhibitor, achieves IC50 = 2.4 nM with an estimated Ki 200 times more potent than the ARA peptide.

Specifications

Chemical Name or Material MM-102 TFA
CAS 1883545-52-5
Color White
Purity Grade Notes Research
Molecular Formula C37H50F5N7O6
Quantity 5 mg
Synonym HMTase Inhibitor IX TFA
Solubility Information DMSO : ≥ 100 mg/mL (127.58 mM)
SMILES O=C(C1(NC([C@@H](NC(C(NC(C(C)C)=O)(CC)CC)=O)CCCNC(N)=N)=O)CCCC1)NC(C2=CC=C(F)C=C2)C3=CC=C(F)C=C3.FC(F)(F)C(O)=O
Molecular Weight (g/mol) 783.83
Formula Weight 783.83
Percent Purity 98.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.