missing translation for 'onlineSavingsMsg'
Learn More

MLN0905, MedChemExpress

Product Code. 16496328
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496328 10 mg 10mg
16458917 5 mg 5mg
16432447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496328 Supplier Medchem Express Supplier No. HY15155

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MLN0905 is a potent PLK1 inhibitor, with an IC50 of 2 nM.

MLN0905 is a potent PLK1 inhibitor, with an IC50 of 2 nM.

Specifications

Chemical Name or Material MLN0905
CAS 1228960-69-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H25F3N6S
Quantity 10 mg
Synonym PLK1 Inhibitor
Solubility Information DMSO : ≥ 30 mg/mL (61.66 mM)
SMILES CC1=NC=C(CCCN(C)C)C=C1NC2=NC(C(C=CC(C(F)(F)F)=C3)=C3NC(C4)=S)=C4C=N2
Molecular Weight (g/mol) 486.56
Formula Weight 486.56
Percent Purity 95.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.