missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ML241 hydrochloride, MedChemExpress
Description
ML241 hydrochloride is a potent p97 inhibitor, inhibiting p97 ATPase with IC50 value of 100 nM.
Specifications
Specifications
| Chemical Name or Material | ML241 hydrochloride |
| CAS | 2070015-13-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H25ClN4O |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 34 mg/mL (83.15 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | Cl[H].C1(N2C3=CC=CC=C3OCC2)=NC(NCC4=CC=CC=C4)=C5CCCCC5=N1 |
| Molecular Weight (g/mol) | 408.92 |
| Formula Weight | 408.92 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?