missing translation for 'onlineSavingsMsg'
Learn More

ML241 hydrochloride, MedChemExpress

Product Code. 16417726
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417726 100 mg 100mg
16487908 10 mg 10mg
16443857 50 mg 50mg
16430378 5 mg 5mg
16423696 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16417726 Supplier Medchem Express Supplier No. HY19797A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML241 hydrochloride is a potent p97 inhibitor, inhibiting p97 ATPase with IC50 value of 100 nM.

ML241 hydrochloride is a potent p97 inhibitor, inhibiting p97 ATPase with IC50 value of 100 nM.

Specifications

Chemical Name or Material ML241 hydrochloride
CAS 2070015-13-1
Color White
Purity Grade Notes Research
Molecular Formula C23H25ClN4O
Quantity 100 mg
Solubility Information DMSO : ≥ 34 mg/mL (83.15 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES Cl[H].C1(N2C3=CC=CC=C3OCC2)=NC(NCC4=CC=CC=C4)=C5CCCCC5=N1
Molecular Weight (g/mol) 408.92
Formula Weight 408.92
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.